ENA238713534 Enamine

4-(1-bromoethyl)-2-chloro-1-fluorobenzene

  • MDL number MFCD11180334
  • Cas Number : 1094438-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-52231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52231
Availability details for
Quantity
  • Availability
  • Add to Cart