ENA238713538 Enamine

5-(chloromethyl)-1-[4-(difluoromethoxy)phenyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD11185601
  • Cas Number : 1094370-85-0
  • Molecular Weight : 260.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52237
Availability details for
Quantity
  • Availability
  • Add to Cart