ENA238713546 Enamine

3-chloro-N-cyclopentylpropanamide

  • MDL number MFCD02973947
  • Cas Number : 349403-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52255

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52255
Availability details for
Quantity
  • Availability
  • Add to Cart