ENA238713547 Enamine

4-[(4-chlorophenyl)sulfanyl]aniline

  • MDL number MFCD00093472
  • Cas Number : 32631-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52256
Availability details for
Quantity
  • Availability
  • Add to Cart