ENA238713551 Enamine

{[(thiophen-2-yl)methylidene]amino}thiourea

  • MDL number MFCD23698441
  • Cas Number : 1021928-63-1
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52261
Availability details for
Quantity
  • Availability
  • Add to Cart