ENA238713552 Enamine

4-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD11209017
  • Cas Number : 261528-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52262

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-52262
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart