ENA238713558 Enamine

2-(3-fluorophenoxy)aniline

  • MDL number MFCD04971079
  • Cas Number : 640766-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52270
Availability details for
Quantity
  • Availability
  • Add to Cart