ENA238713559 Enamine

1-(aminomethyl)-N,N,3-trimethylcyclohexan-1-amine

  • MDL number MFCD09946464
  • Cas Number : 1017057-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52271
Availability details for
Quantity
  • Availability
  • Add to Cart