ENA238713560 Enamine

4-butoxy-2-methylaniline

  • MDL number MFCD08687155
  • Cas Number : 150748-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52272
Availability details for
Quantity
  • Availability
  • Add to Cart