ENA238713562 Enamine

3-bromo-N-[2-(diethylamino)ethyl]aniline

  • MDL number MFCD11153147
  • Cas Number : 52158-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52274
Availability details for
Quantity
  • Availability
  • Add to Cart