ENA238713565 Enamine

2-(1-bromoethyl)-1,4-difluorobenzene

  • MDL number MFCD11180290
  • Cas Number : 655-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-52091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52091
Availability details for
Quantity
  • Availability
  • Add to Cart