ENA238713568 Enamine

1-(4-methylphenyl)-2-(2,2,2-trifluoroethoxy)ethan-1-one

  • MDL number MFCD12062607
  • Cas Number : 1157999-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52289
Availability details for
Quantity
  • Availability
  • Add to Cart