ENA238713578 Enamine

2,2-dimethyl-3,4-dihydro-2H-1-benzopyran

  • MDL number MFCD00052285
  • Cas Number : 1198-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52303

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52303
Availability details for
Quantity
  • Availability
  • Add to Cart