ENA238713582 Enamine

[4-bromo-2-(dimethylamino)phenyl]methanol

  • MDL number MFCD11651979
  • Cas Number : 1099693-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52308
Availability details for
Quantity
  • Availability
  • Add to Cart