ENA238713589 Enamine

2-[(thiophen-3-yl)formamido]acetic acid

  • MDL number MFCD09049197
  • Cas Number : 72196-45-3
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52322
Availability details for
Quantity
  • Availability
  • Add to Cart