ENA238713590 Enamine

3-(4-methoxyphenyl)propanehydrazide

  • MDL number MFCD00463001
  • Cas Number : 121670-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52323
Availability details for
Quantity
  • Availability
  • Add to Cart