ENA238713596 Enamine

methyl 4-(sulfamoylmethyl)benzoate

  • MDL number MFCD11649751
  • Cas Number : 110654-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52331
Availability details for
Quantity
  • Availability
  • Add to Cart