ENA238713597 Enamine

2-methyl-4-(piperidin-4-yl)morpholine

  • MDL number MFCD09941106
  • Cas Number : 1016841-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52094
Availability details for
Quantity
  • Availability
  • Add to Cart