ENA238713600 Enamine

3-(propan-2-yloxy)benzaldehyde

  • MDL number MFCD01922005
  • Cas Number : 75792-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52339
Availability details for
Quantity
  • Availability
  • Add to Cart