ENA238713608 Enamine

2-(3-fluoro-4-methoxyphenyl)oxirane

  • MDL number MFCD09937964
  • Cas Number : 1016806-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52347
Availability details for
Quantity
  • Availability
  • Add to Cart