ENA238713620 Enamine

2-{[(oxolan-2-yl)methyl]sulfanyl}propanoic acid

  • MDL number MFCD09929367
  • Cas Number : 1016754-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52371
Availability details for
Quantity
  • Availability
  • Add to Cart