ENA238713626 Enamine

2-[2-(1H-1,2,4-triazol-1-yl)acetamido]acetic acid

  • MDL number MFCD11172669
  • Cas Number : 1042636-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52383
Availability details for
Quantity
  • Availability
  • Add to Cart