ENA238713627 Enamine

2-chloro-3-(3-methyl-1,2,4-oxadiazol-5-yl)pyridine

  • MDL number MFCD08273492
  • Cas Number : 890095-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52385
Availability details for
Quantity
  • Availability
  • Add to Cart