ENA238713639 Enamine

2-cyclohexylideneacetic acid

  • MDL number MFCD01815140
  • Cas Number : 1552-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52411
Availability details for
Quantity
  • Availability
  • Add to Cart