ENA238713661 Enamine

1-(3-chloro-4-fluorophenyl)ethan-1-ol

  • MDL number MFCD04973766
  • Cas Number : 878572-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52456
Availability details for
Quantity
  • Availability
  • Add to Cart