ENA238713666 Enamine

4-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD00049345
  • Cas Number : 4771-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52466

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52466
Availability details for
Quantity
  • Availability
  • Add to Cart