ENA238713668 Enamine

2-[(dimethylamino)methyl]benzoic acid hydrochloride

  • MDL number MFCD11841286
  • Cas Number : 90279-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52119
Availability details for
Quantity
  • Availability
  • Add to Cart