ENA238713700 Enamine

1-(4-bromophenyl)pentane-1,3-dione

  • MDL number MFCD11185177
  • Cas Number : 960127-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52534
Availability details for
Quantity
  • Availability
  • Add to Cart