ENA238713701 Enamine

4-(3-methylphenyl)butan-2-one

  • MDL number MFCD11655136
  • Cas Number : 102611-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52537
Availability details for
Quantity
  • Availability
  • Add to Cart