ENA238713716 Enamine

2-(4-acetylpiperazin-1-yl)benzaldehyde

  • MDL number MFCD11156387
  • Cas Number : 1021240-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52568
Availability details for
Quantity
  • Availability
  • Add to Cart