ENA238713720 Enamine

[(4-bromophenyl)methyl][2-(dimethylamino)ethyl]amine

  • MDL number MFCD07408130
  • Cas Number : 99862-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52572
Availability details for
Quantity
  • Availability
  • Add to Cart