ENA238713726 Enamine

3-(trifluoromethyl)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD00179830
  • Cas Number : 62980-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52142
Availability details for
Quantity
  • Availability
  • Add to Cart