ENA238713728 Enamine

4-(4-methylpiperazine-1-carbonyl)aniline

  • MDL number MFCD00973892
  • Cas Number : 55121-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52143
Availability details for
Quantity
  • Availability
  • Add to Cart