ENA238713734 Enamine

1-(3-chlorobenzoyl)-1,4-diazepane

  • MDL number MFCD05879261
  • Cas Number : 815651-32-2
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52592
Availability details for
Quantity
  • Availability
  • Add to Cart