ENA238713736 Enamine

1-amino-N-(propan-2-yl)cyclohexane-1-carboxamide

  • MDL number MFCD09938693
  • Cas Number : 1017019-29-2
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52594
Availability details for
Quantity
  • Availability
  • Add to Cart