ENA238713739 Enamine

3-(naphthalene-2-sulfonamido)benzoic acid

  • MDL number MFCD02361166
  • Cas Number : 186032-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52598
Availability details for
Quantity
  • Availability
  • Add to Cart