ENA238713740 Enamine

N'-[(3Z)-1-methyl-2-oxo-2,3-dihydro-1H-indol-3-ylidene]pyridine-3-carbohydrazide

  • MDL number MFCD00565855
  • Cas Number : 341983-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52599

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52599
Availability details for
Quantity
  • Availability
  • Add to Cart