ENA238713745 Enamine

N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide

  • Cas Number : 1055926-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52146
Availability details for
Quantity
  • Availability
  • Add to Cart