ENA238713756 Enamine

4-methyl-N-(2,2,2-trifluoroethyl)-1,3-thiazol-2-amine

  • MDL number MFCD11119543
  • Cas Number : 1036482-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52063
Availability details for
Quantity
  • Availability
  • Add to Cart