ENA238713764 Enamine

3-(prop-2-en-1-yloxy)aniline

  • MDL number MFCD01902259
  • Cas Number : 74900-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52657

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P311,P312,P314,P330,P361,P363,P403+P233,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-52657
pictograms GHS06,GHS08
hstatements H301,H311,H331,H373
Availability details for
Quantity
  • Availability
  • Add to Cart