ENA238713770 Enamine

1-(3,4-dimethoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD01897603
  • Cas Number : 91252-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52693
Availability details for
Quantity
  • Availability
  • Add to Cart