ENA238713789 Enamine

1-(1-bromoethyl)-2,4-dichlorobenzene

  • MDL number MFCD11103119
  • Cas Number : 20444-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-52051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52051
Availability details for
Quantity
  • Availability
  • Add to Cart