ENA238713791 Enamine

5-(chloromethyl)-1-(2,6-dimethylphenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11134282
  • Cas Number : 1094328-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52159
Availability details for
Quantity
  • Availability
  • Add to Cart