ENA238713796 Enamine

[(furan-3-yl)methyl](methyl)amine

  • MDL number MFCD08065551
  • Cas Number : 23008-21-1
  • Molecular Weight : 111.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52163
Availability details for
Quantity
  • Availability
  • Add to Cart