ENA238713801 Enamine

2-(3-amino-1H-pyrazol-1-yl)acetamide

  • MDL number MFCD12149368
  • Cas Number : 1003011-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52167
Availability details for
Quantity
  • Availability
  • Add to Cart