ENA238713808 Enamine

4-(chloromethyl)-2-(phenoxymethyl)-1,3-thiazole

  • MDL number MFCD11178166
  • Cas Number : 1082913-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52180
Availability details for
Quantity
  • Availability
  • Add to Cart