ENA238713809 Enamine

2-(1H-indol-1-yl)acetic acid

  • MDL number MFCD00047262
  • Cas Number : 24297-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52181

Safety Information

signalword Danger
safety_statement 16-27-45
msds https://www.enaminestore.com/catalog/EN300-52181
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart