ENA238713845 Enamine

5-[2-(4-methoxyphenyl)ethyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09802732
  • Cas Number : 1016749-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52200
Availability details for
Quantity
  • Availability
  • Add to Cart