ENA238713847 Enamine

2-(pyrrolidin-1-yl)ethane-1-thiol

  • MDL number MFCD11190758
  • Cas Number : 42302-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52059
Availability details for
Quantity
  • Availability
  • Add to Cart