ENA238713850 Enamine

5-(chloromethyl)-1-(4-methanesulfonylphenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11134271
  • Cas Number : 1094333-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52206
Availability details for
Quantity
  • Availability
  • Add to Cart